missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell Safety Products™ Polytril™ Air 3/4 Gloves
Click to view available options
Size:
7
8
Description
ul>Black, non-pilling polyamide knitted glove suitable for precision work. Suitable for handling fine products or dirty objects in greasy, damp or dirty environments. Excellent abrasion and tear resistance. Black nitrile foam coating on palm and fingers provides oil impermeability, abrasion resistance, and good grip. Tight knit provides comfort, support, dexterity, touch and limits dirt penetration. Easily laundered for extended use. Ventilated back optimizes breathability. Elastic wrist. Black color suitable for work in soiled environments.
Specifications
Specifications
Certifications/Compliance | CE CATEGORY II Intermediary risks, EN 420-2003 Basic standards, EN 388-2003 Mechanical hazard, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number N°0075/797/162/05/07/0193, EC Attestation Number N°0075/797/162/05/07/0193 |
Classification | Dexterity: 5 |
Coated Area | Palm, Fingers |
Construction | Seamless Knit |
Description | Gauge 13 |
Coating Material | Nitrile Foam |
Color | Black |
Cut Resistance Rating | 1 |
Material | Nitrile, Polyamide |
Product Type | Gloves |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'