Learn More
Kinematica™ Polytron™ Homogenizer Stands: Safety Rings
Prevents homogenizer from moving out of position.
27.00 € - 54.00 €
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
10002153
|
Kinematica™
11045035 |
Safety Positioning Ring | ST-P10/600 stand |
54.00 €
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
10233152
|
Kinematica™
11045031 |
Safety Positioning Ring | ST-F20/600, PT-ST 2101, PT-ST 4001 dispersion units |
27.00 €
Each |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||||
Description
The safety ring prevents uncontrolled lowering of the drive unit. Can also be used to fixing the drive unit height.Specifications
Safety Positioning Ring | |
Stainless Steel |