missing translation for 'onlineSavingsMsg'
Learn More

New Pig™ Polyurethane Drain Cover

Encompass_Preferred

274.84 € - 557.52 €

Specifications

Color Yellow
Material Polyurethane
Type Drain Cover
Packaging Quantity 1 Ea.
Height (Metric) 1.1 mm
View More Specs

Products 2
Product Code Brand Type Width (Metric) Price Quantity & Availability  
Product Code Brand Type Width (Metric) Price Quantity & Availability  
12923635
Encompass_Preferred
View Documents
New Pig™
PLRE244
Drain Cover 61 mm
274.84 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
12943635
Encompass_Preferred
View Documents
New Pig™
PLRE242
Drain Cover 91 mm
557.52 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Yellow
Drain Cover
1.1 mm
Polyurethane
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
New Pig™ Polyurethane Drain Cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.