missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Stick with Buffered Peptone Water
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
Brand: LABPLAS KSS-67915-BPW
This item is not returnable.
View return policy
Description
- Polyurethane Sani-Stick
- Saturated with buffered peptone water
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile | |
Gloves |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction