missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LABPLAS Polyurethane Sani-Stick with Letheen Broth
Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
271.00 € - 326.00 €
Specifications
Product Type | Hygiene Sponge Sampling Kit |
---|---|
Media Type | Neutralizers |
Sterility | Sterile |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
17906509
|
LABPLAS
KSS-67910-LT |
None |
271.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17916689
|
LABPLAS
KSS-67915-LT |
Gloves |
326.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Polyurethane Sani-Stick
- Saturated with Letheen broth
- Dimensions: 160 x 229 mm
- Holds: 600 mL
- Option to include gloves with kit
Specifications
Hygiene Sponge Sampling Kit | |
Neutralizers | |
Sterile |