missing translation for 'onlineSavingsMsg'
Learn More

LABPLAS Polyurethane Sani-Stick with Neutralizing Buffer

Synthetic polyurethane sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.

271.00 € - 326.00 €

Specifications

Product Type Hygiene Sponge Sampling Kit
View More Specs

Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
17916509
View Documents
LABPLAS
KSS-67910-NE
None
271.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
17946689
View Documents
LABPLAS
KSS-67915-NE
Gloves
326.00 €
Pack of 100
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Polyurethane Sani-Stick
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications

Specifications

Hygiene Sponge Sampling Kit
Videos
SDS
Documents

Documents

Product Certifications