Learn More
Kautex™ PVC Wide Neck Square Container, Transparent, Without Closure
Fill and clean these wide neck PVC containers with ease due to the roomy neck configuration. Supplied without closure.
Brand:  Kautex™ 2000071082
 View more versions of this product
Product Code. 15981411
  • 354.00 € / Pack of 114
Explore more special offers
Description
Description
  • Member of Series 310 container family
  • Closure available separately
TRUSTED_SUSTAINABILITY
Specifications
Specifications
750 mL
69 mm
PVC
114/Cs.
Square
Wide Neck Container
Transparent
Packaging, Dispensing, Sending Samples, Storage, Mailing
114
No
120 mm
Closure, Foam Insert