missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinylpyrrolidone-iodine complex, Thermo Scientific Chemicals
96.08 € - 1010.03 €
Chemical Identifiers
CAS | 25655-41-8 |
---|---|
Molecular Formula | (C6H9NO)nI2 |
Molecular Weight (g/mol) | 364.95 |
MDL Number | MFCD00084483 |
InChI Key | CPKVUHPKYQGHMW-UHFFFAOYSA-N |
Synonym | povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex |
PubChem CID | 410087 |
IUPAC Name | 1-ethenylpyrrolidin-2-one;molecular iodine |
SMILES | II.*-CC(-*)N1CCCC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10105000
|
Thermo Scientific Acros
229471000 |
100 g |
96.08 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10730575
|
Thermo Scientific Acros
229475000 |
500 g |
307.53 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10474905
|
Thermo Scientific Acros
229470025 |
2.5 kg |
1010.03 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25655-41-8 | |
364.95 | |
CPKVUHPKYQGHMW-UHFFFAOYSA-N | |
410087 | |
II.*-CC(-*)N1CCCC1=O |
(C6H9NO)nI2 | |
MFCD00084483 | |
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex | |
1-ethenylpyrrolidin-2-one;molecular iodine |
Specifications
25655-41-8 | |
Brown-Red to Yellow | |
Authentic | |
(C6H9NO)nI2 | |
MFCD00084483 | |
povidone iodine, povidone-iodine, betadine, isodine, pvp iodine, pvp-iodine, pvp-i, isobetadyne, bridine, disphex | |
CPKVUHPKYQGHMW-UHFFFAOYSA-N | |
1-ethenylpyrrolidin-2-one;molecular iodine | |
410087 | |
Powder |
300°C | |
8% max. (5 g, 105°C) | |
Plastic bottle | |
(C6H9NO)n·xI | |
15, 7815 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,isopropanol,glycerol;,practically insoluble in acetone | |
II.*-CC(-*)N1CCCC1=O | |
364.95 | |
9.0 to 12.0% (available iodine) (on dried substance) | |
Polyvinylpyrrolidone-iodine complex |
Safety and Handling
missing translation for 'rtecsNumber' : TR1579600
missing translation for 'tsca' : TSCA
Product Identifier
- Polyvinylpyrrolidone-iodine complex
RUO – Research Use Only