missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Polywipe-C Wipes
BIOPHARMAGREENER_CHOICE

Reduce solvent usage and maximize process control and consistency in general purpose and critical cleaning applications. Contec™ Polywipe-C Wipes are saturated with 100% IPA or a 70% isopropanol/30% deionized water solution.

740.00 € - 1195.00 €

Specifications

Product Type Wipe
Autoclavable Not Autoclavable
Disposable Single-use
Ply 1-ply
View More Specs

Products 2
Product Code Brand Dimensions (L x W) Saturation Solution Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Saturation Solution Price Quantity & Availability  
17284523
BIOPHARMA
View Documents
Contec™
PSCS0012-BPR
30 x 30 cm (12 x 12 in.) 70% IPA/30% DI Water
1195.00 €
Pack of 480
Please to purchase this item. Need a web account? Register with us today!
16655534
GREENER_CHOICE
View Documents
Contec™
PSCS0012
30 x 30 cm (12 x 12 in.) 70% IPA/30% DI water This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Lightweight wipes have laser sealed edges and are laundered and saturated in Contec's ISO Class 4 environment for the lowest levels of particles, fibers and extractable residue
  • Textured wipe is effective, yet cleans surfaces without scratching
  • Made from 100% textured knitted polyester
  • Saturated with 100% IPA or a 70% isopropanol/30% deionized water solution
  • Reduce VOCs associated with IPA
  • Resealable pouch preserves cleanliness and solvent saturation levels
Specifications

Specifications

Wipe
Not Autoclavable
Single-use
1-ply
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.