Learn More
Contec™ Polywipe-C Wipes
Maintain cleanliness in ISO Class 5 cleanrooms. Contec™ Polywipe-C Wipers are a cost-effective upgrade from less critical wipes. A soft, nonabrasive texture cleans surfaces without scratching.
548.41 €
Specifications
Product Type | Cleanroom Wipe |
---|---|
Material | Polyester |
For Use With (Application) | Cleanroom |
Description
- The lightweight Polywipe-C wipe is designed for general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M-3.5) environments
- Ideal for wiping interior surfaces of process tools and other equipment
- A cost-effective upgrade from less critical wipes
- Made with 100% polyester interlock knit for good solvent sorbency and abrasion and chemical resistance
- Textured surface cleans without scratching
- Laundered and packaged in a ISO Class 4 (FED-STD-209E Class 100/M-2.5) cleanroom
- Lightweight 100% polyester interlock knit wiper offers good sorbency with solvents
- Ideal for cleaning interior surfaces of process tools and other equipment
- Soft, nonabrasive texture cleans surfaces without scratching
Specifications
Cleanroom Wipe | |
Polyester | |
Cleanroom |