Learn More
Cytiva POM/EPDM Packing Connectors
Column connectors for use in gel filtraion chromatography applications
112.00 €
Specifications
Type Packing Connector
Temperature (Metric) Max 60°C
Material EPDM, POM
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10392730
View Documents
Cytiva
18-1153-44
For use with XK 16 columns
112.00 €
Each
In Stock
to see stock.
Add to basket
10630581
View Documents
Cytiva
18-1153-45
For use with XK 26 columns
112.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
This packaging connector used to connect another column, being used as a packing reservoir, to a XK 16 or XK 26 column to achieve efficient packing. A properly packed media bed is essential for obtaining optimal results in all forms of liquid chromatography, particularly in gel filtration. The packing connector makes it easy to obtain a well-packed bed of gel filtration and other media in XK 16 and XK 26 Columns, and is especially convenient for fast and easy packing of affinity, IEX, and HIC media to short bed heights.
Specifications
Specifications
Packing Connector
60°C
EPDM, POM