Learn More
Thermo Scientific™ Ponceau S Staining Solution
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot.
Brand:  Thermo Scientific™ A40000278X3
 View more versions of this product
Product Code. 30088667
  • 249.00 € / Pack of 3
Explore more special offers
Description
Description
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Ponceaus S Staining Solution is a ready-to-use membrane stain for evaluating the transfer efficiency of a western blot. This stain is compatible with nitrocellulose and PVDF membranes. Ponceau S staining is reversible and can be removed with a short incubation in 0.1% NaOH.
Features include:
• A nondestructive, reversible method to stain and detect proteins on nitrocellulose and PVDF membranes
• Minimal nonspecific interactions with the membrane surface; provides a reliable method for visualization of protein transfer to a membrane
• Simple and quick staining protocol that results in reddish-pink bands that are easily photographed
• Reversed easily in less than five minutes
• Subsequent western blot detection unaffected as the stain does not alter the membrane-bound protein
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ponceau S Staining Solution, 3 x 1000 mL
0.1% (w/v) in 5% acetic acid
3 x 1000 mL
Colorimetric
Western Blotting