missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ POP-6™ Polymer for 3500 Dx/3500xL Dx Genetic Analyzers

POP polymers are pre-formulated in a single-use pouch, which not only saves time but also ensures reliability and consistency.

326.00 € - 764.00 €

Specifications

Capillary Length 50 cm, 36 cm
Content And Storage Store the polymer pouch at 2-8°C until ready to use.
Note that expired pouches cannot be used on the instrument.

On-instrument life⁄usage

3500 Dx (8 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 60 injections

3500xL Dx (24 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 20 injections

The pouch includes additional polymer volume to accommodate both installation and a reasonable number of post-installation wizard operations such as bubble removal without affecting the number of samples⁄injections available.
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
For Use With (Application) Fragment Analysis, Standard Sequencing
Product Line POP-6™
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15908826
View Documents Promotion Details
Applied Biosystems™
4393716
384 Samples
326.00 €
384 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15988816
View Documents Promotion Details
Applied Biosystems™
4393711
960 Samples
764.00 €
960 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3500 Dx Series POP-6™ Polymer is a separation matrix for the 3500 Dx/3500xL Dx genetic analyzers. It is conveniently offered in easy-to-use pouch packages in either 960- or 384-sample size formats.

Order Info

Each

Specifications

Specifications

50 cm, 36 cm
3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
POP-6™
2 hrs for 50 cm capillary (long read), 1 hr for 36 cm capillary (std. seq.)
Store the polymer pouch at 2-8°C until ready to use.
Note that expired pouches cannot be used on the instrument.

On-instrument life⁄usage

3500 Dx (8 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 60 injections

3500xL Dx (24 capillary) Genetic Analyzer: lesser of 7 days or 384 samples or 20 injections

The pouch includes additional polymer volume to accommodate both installation and a reasonable number of post-installation wizard operations such as bubble removal without affecting the number of samples⁄injections available.
Fragment Analysis, Standard Sequencing
Wet Ice
Videos
SDS
Documents

Documents

Product Certifications

For in-vitro diagnostic use