Learn More
Applied Biosystems™ POP-7™ Polymer, for 3500/SeqStudio™ Flex
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
78.50 € - 311.00 €
Specifications
For Use With (Application) | Long-Read Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing) |
---|---|
Product Line | POP-7™ |
Description
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages, POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis. POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
POP-4, POP-6, and POP-7 are conveniently offered in easy to use pouch packages with multiple sample size formats. The POP-7 separation matrix is optimized for short- to long-read sequencing and fragment analysis.
• Easy to use: pre-formulated and in a single use pouch, which not only saves time but also ensures reliability and consistency
• Include radio frequency identification (RFID) label: easy to track important information such as part⁄lot number, samples remaining, and expiry dates (displayed on software dashboard)
• Robust formulation: formulated for sequencing and fragment analysis applications
• Replenishable: allow capillaries to be used multiple times
Reproducible Results
POP polymers dynamically coat the capillary wall to control electro-osmotic flow. Their definite quality and uniform consistency eliminate guesswork and help to ensure reproducibility. POP Polymers are specifically formulated to separate DNA fragments of a known size range at a desired resolution and run time.
Many Choices
POP-7 polymers are very versatile and designed for a wide range of short read to long read sequencing applications as well as fragment analysis. POP-4 polymers are designed specifically for HID⁄Forensic applications, while POP-6 polymers are the choice for certain sequencing applications (and fragment analysis).
For partially used pouches that need to be stored, a cap (4412619) that plugs into the pouch fitment opening is recommended.
Order Info
Each
Specifications
Long-Read Sequencing, Short Read Sequencing, Fragment Analysis (Denaturing) | |
POP-7™ |
Safety and Handling
Recommended Storage : The pouch contains 96 samples of the polymer. Store pouch at 2–8°C until ready to use. Refer to the expiration date on the label. Note that expired pouches should not be used on the instrument.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.