Learn More
Haldenwanger™ Porcelain Buchner Funnel
Use these funnels for a variety of filtration applications. Haldenwanger™ Porcelain Buchner Funnels are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 127C/8
 View more versions of this product
Product Code. 10579021
  • 1435.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • Available with perforated fixed filter plate
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
10,600 mL
350 mm
330 mm
Buchner Funnel
Porcelain
330 mm
1Each