Learn More
Haldenwanger™ Porcelain Buchner Funnel
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
93.39 € - 264.00 €
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Plate | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Plate | Price | Quantity & Availability | |||||
10433281
|
Haldenwanger™
L 127C/0A |
50 mL | 48mm |
93.39 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
|||||
10343221
|
Haldenwanger™
L 127C/1 |
75 mL | 59mm |
101.71 €
Each |
In Stock
Log in to see stock. ![]() |
|||||
10251201
|
Haldenwanger™
L 127C/2A |
290 mL | 90mm |
157.00 €
Each |
Estimated Shipment: 09-06-2025
Log in to see stock. |
|||||
10179630
|
Haldenwanger™
L 127C/3 |
580 mL | 114mm |
251.00 €
Each |
Estimated Shipment: 03-06-2025
Log in to see stock. |
|||||
10771752
|
Haldenwanger™
L 127C/2 |
135 mL | 74mm |
264.00 €
Each |
Estimated Shipment: 23-05-2025
Log in to see stock. |
|||||
10434431
|
Haldenwanger™
L 127C/4 |
795 mL | 129mm |
229.00 €
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
|||||
Description
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
- Vertical sides
- Available in a variety of sizes to fit many filtration needs
- Perforated fixed filter plate
Specifications
Porcelain | |
1Pack | |
Buchner Funnel |