Learn More
Haldenwanger™ Porcelain Crucible Lids
Use these lids with porcelain crucibles. Haldenwanger™ Porcelain Crucible Lids are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 79D/1
 View more versions of this product
Product Code. 12807703
  • 95.20 € / Pack of 5
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
87mm
Crucible Lid