Learn More
DWK Life Sciences Porcelain Desiccator Plate
The desiccator insert is a sieve plate on which the substance to be dried is placed in an appropriate container.
Brand:  DWK Life Sciences 297254608
 View more versions of this product
Product Code. 10020091
  • 39.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
The desiccator insert is a sieve plate that separates the desiccator base into two areas - there is a desiccant in the lower part and the substance to be dried is placed in the upper part. The substance to be dried is placed on the sieve plate in an appropriate container for drying.
  • The heat-resistant material allows hot samples to be placed into the desiccator
  • Made of heat-resistant material
TRUSTED_SUSTAINABILITY
Specifications
Specifications
90 mm
Porcelain
Drying of materials
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DWK Life Sciences Porcelain Desiccator Plate > DN 100