Learn More
Haldenwanger™ Porcelain Desiccator Plates
Use these plates for a variety of applications. Haldenwanger™ Porcelain Desiccator Plates feature a smooth internal finish, constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ 119C/280
 View more versions of this product
Product Code. 10241481
  • 205.00 € / Each
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
280 mm
1 Each
Desiccator Plate
Porcelain