Learn More
Haldenwanger™ Porcelain Evaporating Dishes
Designed with glazed laboratory porcelain to provide excellent thermal and chemical resistance. Haldenwanger™ Porcelain Evaporating Dishes provide a solid basin for evaporating excessive solvents to produce a concentrated solution or solid precipitate.
9.01 € - 34.50 €
Specifications
Material Porcelain
Shape Round
Type Evaporating Dish
For Use With (Application) Laboratories
Interior Finish Glazed
View More Specs
Products 6
Product Code Brand Bottom Shape Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Bottom Shape Diameter (Metric) Height (Metric) Price Quantity & Availability  
10494531
View Documents
Haldenwanger™
888/4
Flat 100 mm 20 mm
26.15 €
Each
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
10260011
View Documents
Haldenwanger™
888/2
Flat 80 mm 16 mm
19.70 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
10483381
View Documents
Haldenwanger™
109/5
Round 125 mm 50 mm
34.50 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
10269530
View Documents
Haldenwanger™
109/000
Round 63 mm 25 mm
9.01 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
10128160
View Documents
Haldenwanger™
109/00
Round 70 mm 28 mm
13.20 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
10443001
View Documents
Haldenwanger™
109/0
Round 80 mm 32 mm
15.15 €
Each
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
Description
Description
  • Available in shallow and semi-shallow form
  • Sizes up to 175mL glazed; 1,000mL and larger are glazed except outside base
Specifications
Specifications
Porcelain
Evaporating Dish
Glazed
Round
Laboratories