Learn More
Haldenwanger™ Porcelain Hirsch Funnel
With perforated fixed filter plate
Brand:  Haldenwanger™ 126/4/0
 View more versions of this product
Product Code. 10190131
  • 38.37 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
With perforated fixed filter plate.
Specifications
Specifications
Porcelain
11mm
With fixed filter plate
Hirsch Funnel
56 mm
11mm
1Each