Learn More
Haldenwanger™ Porcelain Hirsch Funnel
With perforated fixed filter plate
Brand:  Haldenwanger™ 126/4/0A
 View more versions of this product
Product Code. 12388567
  • 47.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Available in a variety of sizes
  • With perforated fixed filter plate
  • Complies with DIN VDE 0335, type C 100
With perforated fixed filter plate.
Specifications
Specifications
Porcelain
13mm
1Each
13mm
With fixed filter plate
Hirsch Funnel