missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Medium Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Medium Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

5.55 € - 55.00 €

Specifications

Material Porcelain
Product Type Specs Melting Crucible
View More Specs

Products 4
Product Code Brand Capacity (Metric) Finish Price Quantity & Availability  
Product Code Brand Capacity (Metric) Finish Price Quantity & Availability  
10077410
View accessory products View Documents Promotion Details
Haldenwanger™
79MF/1
200 mL Glazed Inside and Outside Except Base
25.35 €
Each
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
10290861
View Documents Promotion Details
Haldenwanger™
L 79MF/3
100 mL Glazed Inside and Outside Except Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10291481
View Documents Promotion Details
Haldenwanger™
L 79MF/8
10 mL Glazed Inside and Outside Except Base This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12326497
View accessory products View Documents Promotion Details
Haldenwanger™
79MF/9
5 mL Glazed
55.00 €
Pack of 10
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
 
Description

Description

  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications

Specifications

Porcelain
Melting Crucible
Videos
SDS
Documents

Documents

Product Certifications