missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Pestle
Use these pestles for a variety of laboratory applications. Haldenwanger™ Porcelain Pestles are constructed from optimized laboratory porcelain.
42.40 € - 81.95 €
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
12383338
|
Haldenwanger™
56/8/glazed |
60mm |
81.95 €
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
11940897
|
Haldenwanger™
56/4 |
180mm |
42.40 €
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
Description
- Available in a variety of sizes
- Grinding surfaces are unglazed
- Complies with DIN VDE 0335, type C 100
Specifications
Pestle | |
Porcelain | |
Pestle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title