missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Porcelain Pestles
Use these porcelain pestles for a wide range of applications. Haldenwanger™ Porcelain Pestles are available glazed or unglazed with head sizes from 24 to 85mm in diameter. Stringent inspections, starting with raw materials and continuing down to finished products, guarantee constant quality.
19.65 € - 42.90 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10405011
|
Haldenwanger™
56/00 |
115mm |
19.65 €
Each |
In Stock
Log in to see stock. |
|||||
10693291
|
Haldenwanger™
56/2 |
150mm |
41.40 €
Each |
In Stock
Log in to see stock. |
|||||
11930897
|
Haldenwanger™
56/3 |
175mm |
42.90 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Grinding surfaces are unglazed
- Excellent chemical resistance
- Laboratory porcelain complies with DIN EN 60672-3,Group C100, Type C110
Specifications
Unglazed | |
Pestle | |
Porcelain |