missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Porcelain Tall Form Melting Crucibles

Use these dishes for a variety of applications. Haldenwanger™ Porcelain Tall Form Melting Crucibles feature a smooth internal finish, constructed from optimized laboratory porcelain.

6.80 € - 21.35 €

Specifications

Material Porcelain
Product Type Specs Melting Crucible
View More Specs

Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10201301
View Documents
Haldenwanger™
79C/4
85 mL
21.35 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
10569601
View Documents
Haldenwanger™
L 79C/0/STEMPE
15 mL
8.10 €
Each
Estimated Shipment: 14-01-2025
to see stock.
Add to basket
 
10323271
View Documents
Haldenwanger™
L 79C/1/STEMPE
24 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Available in a variety of sizes
  • Sizes up to 175mL glazed except rim
  • 1,000mL and larger are glazed except outside base
  • Complies with DIN VDE 0335, type C 100
Specifications

Specifications

Porcelain
Melting Crucible
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.