missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Porcine IL-8 ELISA Kit
Porcine IL-8 quantitates porcine IL-8 in serum, plasma, supernatant.
Brand: Invitrogen™ ES15RBX5
This item is not returnable.
View return policy
Description
Porcine IL-8 quantitates porcine IL-8 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant porcine IL-8.
Interleukin 8 (IL-8, CXCL8) is a 72 amino acid pro-inflammatory factor which belongs to the CXC subfamily of chemokines, and are bound by the cell surface receptors IL-8RA and IL-8RB. IL-8 functions as a chemoattractant and potent angiogenic factor. The expression and secretion of IL-8 can be induced by diverse inflammatory stimuli in many cells, including macrophages and endothelial cells. In endothelial cells, IL-8 is present in storage vesicles called Weibel-Palade bodies. IL-8, first isolated from osteosarcoma cells, contains the ELR-motif (N-terminal Glu-Leu-Arg amino acid sequence) and signals through the CXCR1 and CXCR2 receptors. Previous nomenclature for IL-8 includes neutrophil activating protein 1 (NAP-1), granulocyte chemotactic protein 1 (GCP-1), monocyte-derived neutrophil-activating peptide (MONAP) and protein 3-10C. IL-8 and ten other members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. Cancer studies have demonstrated a role for IL-8 in the angiogenesis and growth of tumours, and IL-8 is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection.Specifications
(Ala-IL-8)77; (Ser-IL-8)72; Alveolar macrophage chemotactic factor I; alveolar macrophage-derived chemotactic factor-I; AMCF-I; AN x 7; beta endothelial cell-derived neutrophil activating peptide; beta-thromboglobulin-like protein; chemokine (C-X-C motif) ligand 8; C-X-C motif chemokine 8; C-X-C motif chemokine ligand 8; CXCL8; emoctakin; GCP/IL-8 protein I; GCP/IL-8 protein II; GCP/IL-8 protein III; GCP/IL-8 protein IV; GCP/IL-8 protein V; GCP/IL-8 protein VI; GCP1; GCP-1; Granulocyte chemotactic protein 1; IL8; IL-8; IL-8(1-77); IL-8(5-77); IL-8(6-77); IL-8(7-77); IL-8(8-77); IL-8(9-77); IL8/NAP1 form I; IL8/NAP1 form II; IL8/NAP1 form III; IL8/NAP1 form IV; IL8/NAP1 form V; IL8/NAP1 form VI; ILN; Interleukin; interleukin 8; Interleukin8; interleukin-8; LECT; LUCT; lung giant cell carcinoma-derived chemotactic protein; lymphocyte derived neutrophil activating peptide; Lymphocyte-derived neutrophil-activating factor; LYNAP; MDNCF; MDNCF-a; MDNCF-b; MDNCF-c; MONAP; Monocyte-derived neutrophil chemotactic factor; Monocyte-derived neutrophil-activating peptide; NAF; NAP1; NAP-1; neutrophil activating peptide 1; neutrophil and T-lymphocyte chemotactic and activating factor; Neutrophil attractant/activation protein 1; neutrophil attractant/activation protein-1; Neutrophil-activating factor; neutrophil-activating peptide 1; Neutrophil-activating protein 1; Permeability factor 1; PF1; Protein 3-10 C; RP11-537A6.8; RPF1; small inducible cytokine subfamily B, member 8; SNX; SYNEXIN; T-cell chemotactic factor; tumor necrosis factor-induced gene 1 | |
10 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CXCL8 | |
<10% | |
HRP | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |
10 to 2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
P26894 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
5 x 96 Tests | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
IL-8 | |
4 hr 45 min |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction