missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Porcine Serum

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Beschreibung
- Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays
- It has earned the trust of researchers with consistent quality and award-winning support
- Helps meet research needs and budget requirements
- Sera Category: Specialty
- Origin: New Zealand
- Age: 1 year or less
Bestellinformation
Shipping condition: Frozen (Dry Ice)
Spezifikation
Spezifikation
| Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
| Species | Swine/Porcine, Porcine |
| Age | 1 year or less |
| Country of Origin | New Zealand |
| Form | Liquid |
| Product Type | Porcine Serum |
| Quantity | 500 mL |
| Serum Treatment | Standard (Sterile-filtered) |
| Sterility | Sterile |
| Product Line | Gibco |
| Mehr anzeigen |
For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction