missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ HQ 50 μm Strong Anion Exchange Resin

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance

Brand:  Thermo Scientific™ 82077

 View more versions of this product

Product Code. 15662135

  • 201.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

For Research Use Only. Not for use in diagnostic procedures.

This item is not returnable. View return policy

Description

Description

POROS™ 50-micron resins are available for low pressure applications, including large scale downstream bioprocessing. POROS HQ is based on a quaternized polyethyleneimine functional group yielding a high capacity, Perfusion Chromatography™ resin designed for the separation and purification of biomolecules.

  • Higher productivity—high throughput and high dynamic capacity associated with Perfusion Chromatography
  • Chemical stability—allows aggressive cleaning and sanitization
  • Enhanced biomolecule access—provided via large pores, ranging between 500-10000 Å
  • Polystyrenedivinylbenzene particles—yield a robust, easily packable matrix
  • Develop better separations methods in a shorter time frame—the speed of Perfusion Chromatography technology reduces weeks of experimentation to only a few hours of work, so you have plenty of time to explore all the variables of your separation

Reduce time-consuming sample prep
You can replace many sample preparation steps with high-speed Perfusion Chromatography technology. For instance, dialysis of large volumes of material can be replaced by high flow rate processing of the dilute sample. Elution in a small volume of new buffer accomplishes both buffer exchange and concentration.

Create novel assays for more efficient analysis
Perfusion Chromatography technology is not limited to standard modes of separation. Both enzymes and affinity ligands can be immobilized on POROS media. By combining rapid on-column protein digestions with rapid on-column immunoassays and chromatographic separations, you can create entirely new assays with unlimited potential.

High capacity, high resolution, high speed
In contrast to conventional chromatography media, POROS Perfusion Chromatography resin particles are engineered to have two discreet classes of pores. Large throughpores allow convection flow to occur through the particles themselves, quickly carrying sample molecules to short diffusive pores inside. By reducing the distance over which diffusion needs to occur, the time required for sample molecules to interact with interior binding sites is also reduced. Diffusion is no longer limiting and flow rates can be dramatically increased—without compromising resolution or capacity. Separations can be achieved at 1,000 to 5,000 cm/hr compared to 50 to 360 cm/hr for conventional resins

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HQ Strong Anion Exchange Resin
Separation and Purification of Biomolecules
10 mL
POROS™
Quaternized Polyethyleneimine
Room Temperature
Quaternized Polyethyleneimine
10 mL
Store in refrigerator (2–8°C).
Poros Bead
Protein
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ405203 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number82077
Lot NumberZG397363 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number82077
Lot NumberYH370929 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number82077
Lot NumberXH356757 Certificate TypeCertificate of Analysis Date30/08/2022 Catalog Number82077
Lot NumberWI333068 Certificate TypeCertificate of Analysis Date24/09/2021 Catalog Number82077
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ POROS™ HQ 50 μm Strong Anion Exchange Resin > 10, 50 HQ

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.