missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ CaptureSelect™ HSA Affinity Column, 4.6 x 100 mm

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates.

Brand:  Thermo Scientific™ 4469170

Additional Details : Weight : 0.16300kg

Product Code. 18476788

  • 3570.00 € / Each
Estimated Shipment: 26-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Analytical groups supporting biologic drug development require rapid, sensitive and precise methods for protein quantitation from cell culture, fermentation harvests, or purification intermediates. It is critical for biopharmaceutical manufacturers to be able to monitor bioreactor product titers to determine when to harvest or abort a reactor cycle.

Poros™ CaptureSelect™ HSA HPLC columns incorporate the advantages of high-speed Poros chromatography resins with the unique selectivity of CaptureSelect™ ligands. The combination allows three minute quantitation of HSA fusion proteins.

Features:

  • Broad utility—POROS™ A, G and CaptureSelect™affinity ligands address most biological drug classes
  • Rapid—POROS™ resins offer fast quantitation or purification of immunoglobulins and fusion proteins
  • Hands-off operation—HPLC method can be automated for streamlined operation
  • Robust—deliver high-performance purification over hundreds of chromatography cycles

High Speed with High Specificity

The combination of CaptureSelect™ HSA affinity ligands when combined with POROS™ 20 μm resin provides a high flow rate, high performance chromatography solution for the rapid quantitation and small-scale purification of HSA Fusion proteins.

POROS™ high performance chromatography allows for the quantitation of product from clarified harvest or partially purified intermediates—typically in under three minutes. Moreover, only minimal sample preparation (0.2 μm filtration) is required, and the eluted product is highly purified and suitable for further analysis including mass spectrometry.

Automation

POROS™ affinity chromatography columns are suitable for use with HPLC systems, which allows the automation of chromatography methods, data collection, and data analysis. In addition, the immobilized affinity ligands enable one method and buffer system to be used across a class of molecules, streamlining analytical approaches.

Multiple Cycles

Proper sample preparation, adherence to column operating instructions, and the use of an optimized column cleaning regime results in strong re-use performance. POROS™ Affinity Chromatography columns can be cycled hundreds of times for cost-effective and reproducible quantitation or purification.

HSA Column Specifications:

  • Protein Binding Capacity: 2.6 mg/mL
  • Species Specificity: Human
  • Column Material: PEEK™ polymer or stainless steel
  • Frit: 2 μm, titanium
  • POROS™ Resin: Cross-linked poly (styrenedivinylbenzene)
  • CaptureSelect™ Ligands: Single-domain monospecific antibody fragment produced in Saccharomyces cerevisiae
Specifications

Specifications

2.6 mg/mL
Up to 5000 cm/hr.
0.16 micron
3.4 mL
HPLC
180 bar
Human
20 μm
1 Column
Column
Column
1.7 mL
Polystyrenedivinylbenzene
Column includes EZ grip fittings and column test certificate

Store at 4°C. DO NOT FREEZE.
1.7 mL
Not High-throughput Compatible (Manual)
Wet Ice
Cell Culture Supernatants
CaptureSelect™, POROS™
HSA, HSA Fusion Proteins
CaptureSelect™ HSA
Affinity, Pre-packed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostics procedures.