Learn More
Thermo Scientific™ POROS™ GoPure™ AAV9 Screening Plate, 96-well, 20 μL
Thermo Scientific™ POROS™ GoPure™ AAV9 96-well screening plates contain POROS CaptureSelect AAV9 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis.
Brand:  Thermo Scientific™ A65810
733.60 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 30131540
  • 917.00 € / Each
Explore more special offers
Description
Description
POROS GoPure AAV9 96-well screening plates contain POROS CaptureSelect AAV9 Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.
These pre-filled screening plates have the following features:
  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins
Specifications
Specifications
800 μL
Resin Slurry
1 Plate
20 μL
50 μm
POROS™ CaptureSelect™
Chromatography
96-well Filter Plate