missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ AAVX Screening Plate, 96-well, 20 μL

Product Code. 30131273 Shop All Thermo Scientific Products
missing translation for 'orderingAttributeHoverText'
Quantity:
1 Plate
missing translation for 'unitSize'
Each
This item is not returnable. View return policy

Product Code. 30131273

missing translation for 'mfr': Thermo Scientific™ A65809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ POROS™ GoPure™ AAVX 96-well screening plates contain POROS CaptureSelect AAVX Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis.

POROS GoPure AAVX 96-well screening plates contain POROS CaptureSelect AAVX Affinity Resin and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.

These pre-filled screening plates have the following features:

  • High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
  • 20 μL per well, helping preserve sample while maximizing data generation
  • Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins

Specifications

Capacity (Metric) 800 μL
Particle Size 50 μm
Form Resin Slurry
Product Line POROS CaptureSelect
Quantity 1 Plate
For Use With (Application) Chromatography
Sample Volume 20 μL
Format 96-well Filter Plate

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.