missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ XQ Strong Anion Exchange Resin

POROS XQ is a high-capacity, high-resolution, salt-tolerant resin that enables >140 mg/mL dynamic binding capacity while delivering exceptional separation performance

Brand:  Thermo Scientific™ 82074

580.78 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15652135

  • 818.00 € / 50mL
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket

Alert:

For Research Use Only. Not for use in diagnostic procedures.

This item is not returnable. View return policy

Description

Description

POROS™ XQ Strong Anion Exchange Resin is a next-generation, high-capacity, high-resolution, salt-tolerant resin that that enables >140 mg/mL dynamic binding capacity in the presence of up to 6.6 mS/cm NaCl, while delivering exceptional separation performance. A major improvement to the performance of traditional anion exchangers, POROS XQ resin sets the standard for strong anion exchange chromatography in both bind & elute or flow-through modes of operation.

  • High dynamic binding capacity leading to reduced column size, smaller footprint, decreased water and buffer usage, and reduced cycling
  • High resolution supporting improved product yield and purity, ultimately improving the cost of goods for the molecule being produced
  • Robust salt tolerance allowing enhanced process design flexibility and streamlining of process flow
  • Low operating back pressure and linear pressure versus flow responses driving flexible scalability
  • Rigid polymeric bead with covalent surface chemistry for easier handling and packing, and superior physical and chemical stability enabling a more robust downstream purification process

Dynamic binding capacity
POROS XQ resin has a high binding capacity of more than 140 mg/mL at 5% breakthrough, over a broad range of process conditions, with superior binding capacity across a broad range of pH and salt conditions. Higher dynamic binding capacity was achieved by optimizing the base bead pore structure and surface chemistry.

High resolution
Using an optimized 50 µm particle size, POROS XQ resin maintains the superior resolving capacity of the POROS resins, overcoming the challenging impurity levels associated with high titer processes. Resolution remains consistent across a wide range of operating linear flow rates.

Robust salt tolerance
POROS XQ resin has high and consistent protein capacity across a broad range of salt concentrations up to 140 mg/mL at 50 mM NaCl (6.6 mS/cm conductivity). The ability to load columns under higher conductivity reduces or helps eliminate the dilution of column feed streams or the removal of unit operations, such as tangential flow filtration.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

XQ Strong Anion Exchange Resin
Poros Bead
Protein
Quaternized Polyethyleneimine
Strong Anion Exchange Chromatography
50 mL
Store in refrigerator (2–8°C).
POROS™
Room Temperature
Quaternized Polyethyleneimine
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.