Learn More
Prestige Medical™ Portable Autoclave Classic Media
Use this autoclave where a cost effective, portable autoclave is needed. Prestige Medical™ Portable Autoclave Classic Media is fully automatic, simply press the button to start. Lightweight and portable, ideal for laboratory use. This 12 liter Classic Media autoclave sterilizes at 121°C.
Brand:  Prestige Medical™ 210048UK
Product Code. 12755355
  • 2012.40 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High quality construction
  • Fully automatic electronic time and temperature control
  • Steam release valve automatically opens to release excess pressure
  • Automatic fault warning light and cut-out if unit boils dry or sterilisation is interrupted
  • Dual interlock handles prevent the lid being opened before the pressure and temperature inside have fallen to safe levels
  • Complete with instrument basket
  • Models 2015 onwards available with optional printer for easy data recording
  • CE marked
  • Comprehensive one year warranty (UK)
  • Interlock safety mechanism, eliminates the risk of super-heated vessels exploding
  • Reliable, special in-built software ensures exact temperatures are reached for the correct duration
  • 9L and 12L models
  • Fits neatly onto the smallest of workbenches For the sterilization of media, liquids and contaminated waste
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Prestige Medical™ Portable Autoclave Classic Media > Capacity: 12L