missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Portable Centrifuge Kit

The Portable Centrifuge Kit offers a convenient way to spin down and prepare samples outside of a conventional laboratory environment.

Brand:  Fisherbrand™ 120320

260.90 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 15817944

  • 344.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Description

Description

Immediate Field Processing:

  • Process samples in un-powered remote areas before deterioration can take place.
  • Allows for immediate on-site testing for faster results.
  • Made of durable ABS plastic for easy clean in field environments.

Easy to Use:

  • Centrifuge is equipped with tool-less SnapSpin rotors for easy change out.
  • Simple operation with lock/unlock activation.

Portable Convenience:

  • All items in kit are housed in a handled Transport Box for portability.
  • The handled box has an air-tight seal for optimal sample protection.

Specially Designed Inserts:

  • Seat the centrifuge, all its accessories, and includes a tube rack.

Package Includes:

  • Mini Centrifuge
    • 6 Place 1.5/2.0 mL SnapSpin tube rotor
    • 16 Place 0. 2 mL SnapSpin tube
    • Rotor (singles or 2 x 8 strip)
    • 6 Tube adapters (0.2 mL)
    • 6 Tube adapters (0.5 mL)
    • 4 O-rings
    • Storage case for rotors and adapters
    • 16 Place tube rack
    • Low voltage, double insulated power adapter with 4 interchangeable plugs
  • Transport Box
    • 16 Place tube rack insert
    • Transport boc dividers
    • Mini Centrifuge cradle
  • Portable 12-volt Centrifuge adapter
  • Five-year Warranty
Specifications

Specifications

2,000 x g
No
0°C to 40°C (32°F to 104°F)
5 Years
6,000 rpm
Yes (6 Place, 1.5/2.0 mL SnapSpin)
Centrifuge Kit
153 x 128 x 104 mm (5.97 x 4.99 x 4.05 in.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.