Learn More
Hughes Portable Emergency Eye/Face/Body Wash
Portable, stainless steel emergency eye, face and body shower.
Brand:  Hughes HEWS0038GV
Product Code. 17752338
  • 1320.00 € / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • This portable 15 liter stainless steel cylinder with Optiflex hand shower effectively washes eyes, face and body delivering up to three minutes of water when pressurised via a Schraeder valve
  • The pressure gauge and dual action safety valve prevent over pressurisation and allow the user to vent the cylinder for cleaning and maintenance
Specifications
Specifications
24 cm
Stainless Steel
Emergency Wash
26.7 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hughes Portable Emergency Eye/Face/Body Wash > Emergency Wash; Material: Stainless Steel