Learn More
OHAUS™ Portable Precision Balance
Multi-Purpose Portable Balances Suitable for Everyday Weighing
Brand:  OHAUS™ 30615581
314.71 EUR valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 17217840
  • 334.00 € / 1 unit
Explore more special offers
Description
Description
  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RS232, USB, or Ethernet (available as accessories)
120 g
0.001 g
0.003 g
NV123
2 sec
10°C to 30°C, 80%RH, non-condensing¶operability guaranteed between 5°C to 40°C
230 x 204 x 104 mm
8 in.
3.7 in.
2.2 lb.
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Backlit LCD
Weighing, Parts Counting, Percent Weighing, Checkweighing
ABS housing, stainless steel pan, transportation lock, slip-resistant and adjustable feet, leveling bubble
0.002 g
50°F to 86°F, 80%RH, non-condensing¶operability guaranteed between 41°F to 104°F
Portable Balances
9.1 in.
4.2 in.
93 mm