Learn More
OHAUS™ Portable Precision Balance
Multi-Purpose Portable Balances Suitable for Everyday Weighing
Brand:  OHAUS™ 30615586
356.17 EUR valid until 2025-12-31
Use promo code "24377" to get your promotional price.
 View more versions of this product
Product Code. 17267840
  • 397.00 € / 1 unit
Explore more special offers
Description
Description
  • Navigator offers essential weighing applications at a competitive price point.
  • Advanced weighing technology offers one-second stabilization for fast and accurate weighing results, improving operator efficiency, productivity and throughput.
  • Overload protection system that can withstand loads up to four times the rated capacity safeguards the balance.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RS232, USB, or Ethernet (available as accessories)
3200 g
0.01 g
0.05 g
NV3202
2.5 sec
10°C to 40°C, 85%RH, non-condensing
230 x 204 x 70 mm
144 mm
7.5 in.
2.8 in.
1 kg
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Backlit LCD
Weighing, Parts Counting, Percent Weighing, Checkweighing
ABS housing, stainless steel pan, transportation lock, slip-resistant and adjustable feet, leveling bubble
0.02 g
50°F to 104°F, 85%RH, non-condensing
Portable Balances
9.1 in.
8 in.
190 mm
2.2 lb.