missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters

Designed to measure pH, conductivity (EC), total dissolved solids (TDS), and temperature with a single probe. Hanna Instruments™ Portable Waterproof pH/EC/TDS Meters are available for high range and low range measurement.

Brand:  Hanna Instruments™ HI991300

Product Code. 10182271

  • 1695.00 € / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

This multi-parameter meter takes readings simply and quickly, and it fits in the palm of your hand. From setup to calibration all operations are performed with just two buttons. It features a clear, extra-large LCD display and easy-access rotary knobs to effect calibration changes. A single pre-amplified probe is used for all parameters.

  • Waterproof and rated for IP 67 conditions
  • Easy to clean and maintain
  • Multi-level LCD display
  • On-screen tutorial messages for calibration and setup
  • Automatic Temperature Compensation
  • Automatic one or two-point pH calibration
  • BEPS: Alerts the user in the event that low battery power could adversely affect the readings
  • Battery % displayed on startup
  • Compact, heavy duty, and waterproof
  • Preprogrammed buffer recognition values, TDS factors and temperature coefficients
Specifications

Specifications

HI 1288 probe, batteries, 1m cable, rugged carrying case and instructions
±0.01
Automatic
0.01
0°C to 60.0°C
0.1°C/F
IP 67
205 g
2 Years (6 Months Probe)
±2% F.S.
HI1288 PVC body, pre-amplified multiparameter probe with internal temperature sensor, DIN connector and 1m cable
0 to 2000ppm (mg/L)
1μS/cm
0 to 14 pH
1.5V (3) AAA, Auto-Off after 8 min. non-use
500 hr.
±0.5°C/±1°F
Low-Range Detection
0 to 14
±1999
32.0°F to 140.0°F
LCD
150 x 80 x 36 mm (5.9 x 3.1 x 1.4 in.)
7.2 oz.
Two-button
PVC
0 to 3999μS/cm
TDS Conversion Factor: 0.45 to 1.00
1ppm (mg/L)
pH: automatic; EC/TDS: automatic with β adjustable from 0.0 to 2.4%/°C
Portable pH/EC/TDS/Temperature Meter
±2% full scale
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers