missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ Position Rack Flask Vessel Type
The racks come in a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates
464.28 € - 1256.10 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16616125
|
Gilson™
152420 |
3.5 mL Vial (10 x 75 mm) |
524.70 €
Pack of 5 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
16626125
|
Gilson™
152422 |
9 mL Vial (13 x 100 mm) |
464.28 €
Pack of 5 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
16636125
|
Gilson™
152424 |
25 mL Vial (18 x 150 mm) |
1256.10 €
Pack of 5 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
16646125
|
Gilson™
152426 |
6.8 mL MiniVial (17 x 55 mm) |
556.50 €
Pack of 5 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
16656125
|
Gilson™
152427 |
20 mL Vial (58 x 57 mm) |
551.20 €
Each |
Estimated Shipment: 17-09-2024
Log in to see stock. |
|||||
Description
- These various racks are used with current and legacy Gilson liquid handlers, ASPEC™ systems, and other instruments
- Flask Vessel Type
Specifications
Flask | |
5 Pk. |