missing translation for 'onlineSavingsMsg'
Learn More

Notrax Posture Mat Classic™ Anti-fatigue Mats for Standing Workers

Product Code. 17742128
Click to view available options
Length (Metric):
60 cm
102 cm
152 cm
177 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17742128

Brand: Notrax 425S2472BL

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Notrax Posture Mat - Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations.

  • Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations
  • The rebound of the foam results in an energizing bounce effect that stimulates automatic balance checks to improve posture and constant micro movements to maintain blood flow
  • Made from a durable closed cell foam that is reputed for its ultimate comfort and longevity properties, the naturally resilient material moulds to the foot to spread the weight and reduce pressure points from the hard floor underneath
  • All four sides have bevelled edges to minimize tripping hazards
  • Loose lay with no installation required

Specifications

Color Black
Length (English) 5.8 ft.
Length (Metric) 177 cm
Material Rubber
Type Industrial and Commercial Matting
Width (Metric) 60 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.