missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Posture Mat Classic™ Anti-fatigue Mats for Standing Workers
Notrax Posture Mat - Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations.
Brand: Notrax 425S3640BL
This item is not returnable.
View return policy
Description
- Exceptionally soft and comfortable anti-fatigue mats for standing workers in primarily static positions with limited lateral movement making it ideal for sit-stand desks and individual standing workstations
- The rebound of the foam results in an energizing bounce effect that stimulates automatic balance checks to improve posture and constant micro movements to maintain blood flow
- Made from a durable closed cell foam that is reputed for its ultimate comfort and longevity properties, the naturally resilient material moulds to the foot to spread the weight and reduce pressure points from the hard floor underneath
- All four sides have bevelled edges to minimize tripping hazards
- Loose lay with no installation required
Specifications
Black | |
102 cm | |
Industrial and Commercial Matting |
3.3 ft. | |
Rubber | |
91 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Notrax Posture Mat Classic™ Anti-fatigue Mats for Standing Workers > Matting; Length: 102 cm; Width: 91 cm
Spot an opportunity for improvement?Share a Content Correction