Learn More
Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate, 95%, Thermo Scientific™
77.80 € - 294.00 €
Chemical Identifiers
CAS | 1150655-10-9 |
---|---|
Molecular Formula | C8H3BF9K |
Molecular Weight (g/mol) | 320.006 |
MDL Number | MFCD07368252 |
InChI Key | OXVQSBLFWHNZJA-UHFFFAOYSA-N |
Synonym | 2,4-bis trifluoromethyl phenyltrifluoroborate potassium salt, potassium 2,4-bis trifluoromethyl phenyltrifluoroborate, potassium 2,4-bis trifluoromethyl phenyl trifluoroboranuide, potassium 2,4-bis trifluoromethyl phenyl trifluoro borate 1-, potassium ion 2,4-bis trifluoromethyl phenyl trifluoroboranuide |
PubChem CID | 44717658 |
IUPAC Name | potassium;[2,4-bis(trifluoromethyl)phenyl]-trifluoroboranuide |
SMILES | [B-](C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1150655-10-9 | |
320.006 | |
OXVQSBLFWHNZJA-UHFFFAOYSA-N | |
44717658 | |
[B-](C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+] |
C8H3BF9K | |
MFCD07368252 | |
2,4-bis trifluoromethyl phenyltrifluoroborate potassium salt, potassium 2,4-bis trifluoromethyl phenyltrifluoroborate, potassium 2,4-bis trifluoromethyl phenyl trifluoroboranuide, potassium 2,4-bis trifluoromethyl phenyl trifluoro borate 1-, potassium ion 2,4-bis trifluoromethyl phenyl trifluoroboranuide | |
potassium;[2,4-bis(trifluoromethyl)phenyl]-trifluoroboranuide |
Specifications
173°C to 178°C | |
Brown to White | |
C8H3BF9K | |
2,4-bis trifluoromethyl phenyltrifluoroborate potassium salt, potassium 2,4-bis trifluoromethyl phenyltrifluoroborate, potassium 2,4-bis trifluoromethyl phenyl trifluoroboranuide, potassium 2,4-bis trifluoromethyl phenyl trifluoro borate 1-, potassium ion 2,4-bis trifluoromethyl phenyl trifluoroboranuide | |
[B-](C1=C(C=C(C=C1)C(F)(F)F)C(F)(F)F)(F)(F)F.[K+] | |
320.006 | |
320.01 | |
Potassium 2,4-bis(trifluoromethyl)phenyltrifluoroborate |
1150655-10-9 | |
Solid | |
MFCD07368252 | |
OXVQSBLFWHNZJA-UHFFFAOYSA-N | |
potassium;[2,4-bis(trifluoromethyl)phenyl]-trifluoroboranuide | |
44717658 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold; Store under Argon