missing translation for 'onlineSavingsMsg'
Learn More

Potassium, 98%, chunks, in mineral oil, Thermo Scientific Chemicals

45.12 € - 722.60 €

Chemical Identifiers

CAS 7440-09-7
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10216760
View Documents
Thermo Scientific Acros
223860100
10 g Plastic Bottle in Tin Can
45.12 €
10g
Please to purchase this item. Need a web account? Register with us today!
10210903
View Documents
Thermo Scientific Acros
223860500
50 g Plastic bottle
126.63 €
50g
Please to purchase this item. Need a web account? Register with us today!
10347021
View Documents
Thermo Scientific Acros
223861000
100 g Plastic bottle
216.08 €
100g
Please to purchase this item. Need a web account? Register with us today!
10369233
View Documents
Thermo Scientific Acros
223865000
500 g Plastic Drum
722.60 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-09-7
39.10
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
5462222
potassium
K
MFCD00133776
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
CHEBI:26216
[K]

Specifications

Gray
98%
K
01,905; 06,274; 12,400; 13,170; 14,308; 15,269
kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
[K]
39.10
CHEBI:26216
Chunk
7440-09-7
K
MFCD00133776
15, 7725
ZLMJMSJWJFRBEC-UHFFFAOYSA-N
potassium
5462222
39.1
Potassium
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

Supplemental information
  • MIXTURE LIST-Contains : potassium

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only