missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Acetate, puriss., Honeywell Fluka™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
63.18 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 kg |
Linear Formula | CH3COOK |
UN Number | NONH for all modes of transport |
Beilstein | 3595449 |
Formula Weight | 98.14g/mol |
Percent Purity | 99 to 101% |
Grade | puriss. |
Density | 1.57g/cm3at 25°C |
Chemical Name or Material | Potassium Acetate, Meets analytical specification of Ph. Eur. BP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'