missing translation for 'onlineSavingsMsg'
Learn More

Potassium acrylate hydrate, 95%, Thermo Scientific Chemicals

Product Code. 11332427
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10192-85-5
Molecular Formula C3H3KO2
Molecular Weight (g/mol) 110.15
MDL Number MFCD00067206
InChI Key ZUBIJGNKOJGGCI-UHFFFAOYSA-M
Synonym potassium acrylate, potassium polyacrylate, 2-propenoic acid, potassium salt, potassium prop-2-enoate, 79-10-7 parent, polyacrylic acid, potassium salt, 2-propenoic acid, homopolymer, potassium salt, acrylate potassium, acrylic acid homopolymer potassium salt, acmc-20ajv9
PubChem CID 4429391
IUPAC Name potassium;prop-2-enoate
SMILES [K+].[O-]C(=O)C=C

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 3695913
Formula Weight 110.16 (Anhydrous)
Percent Purity 95%
Chemical Name or Material Potassium acrylate hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'