missing translation for 'onlineSavingsMsg'
Learn More
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 10174800

Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 10174800
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10174800

Brand: Thermo Scientific Acros 223801000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28300-74-5
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 333.93
MDL Number MFCD00167056
Synonym Antimonyl potassium tartrate sesquihydrate

Specifications

Melting Point 100.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 100 g
Assay Percent Range 99% min. (Iodometry)
Merck Index 15, 691
Solubility Information Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol
Formula Weight 333.93
Percent Purity ≥99%
Infrared Spectrum Authentic
Loss on Drying 2.7% max. (105°C)
Packaging Glass bottle
Chemical Name or Material Potassium antimonyl tartrate sesquihydrate, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468850 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number223805000,223801000,223800025
Lot NumberA0467271 Certificate TypeCertificate of Analysis Date29/10/2024 Catalog Number223805000,223801000,223800025
Lot NumberA0463896 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number223805000,223801000,223800025
Lot NumberA0458628 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number223805000,223801000,223800025
Lot NumberA0438014 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number223805000,223801000,223800025
compliance-icons compliance-icons
Product Identifier
  • Potassium antimonyl tartrate sesquihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash thoroughly after handling.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Potassium antimonyl tartrate trihydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.