missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonyl tartrate sesquihydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 15234684
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 28300-74-5
Melting Point 100.0°C
Packaging Glass bottle
Molecular Formula 1·5 H2O
Quantity 100 g
Merck Index 15691
Synonym Antimonyl potassium tartrate sesquihydrate
Solubility Information Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water,1g/15mL glycerol,insoluble in alcohol
Molecular Weight (g/mol) 333.93
Formula Weight 333.93
Grade ACS Reagent
Chemical Name or Material Potassium antimonyl tartrate sesquihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'