missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Honeywell Fluka™
38.39 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
1 kg
25 kg
6 x 1 kg
Packaging:
Paper / Poly Bag
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 1 kg |
Linear Formula | KHCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4535309 |
Solubility Information | Soluble in water |
Formula Weight | 100.12g/mol |
Percent Purity | 99.5 to 101% (acidimetric) |
Grade | puriss. |
Packaging | Poly bottle |
Chemical Name or Material | Potassium Bicarbonate, Meets analytical specification of Ph. Eur. BP, USP |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'