missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate USP/FCC Powder, Macron Fine Chemicals™
Click to view available options
Quantity:
500 g
Packaging:
POLYSTORMOR
Specifications
Specifications
CAS | 298-14-6 |
Physical Form | Powder |
Quantity | 500 g |
Molecular Formula | CHKO3 |
MDL Number | MFCD00011402 |
Synonym | potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 |
InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
SMILES | [K+].OC([O-])=O |
IUPAC Name | potassium;hydrogen carbonate |
Molecular Weight (g/mol) | 100.11 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction